missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl Succinate, TRC
Specifications
Specifications
| Chemical Name or Material | Monomethyl Succinate |
| CAS | 3878-55-5 |
| Molecular Formula | C5H8O4 |
| Quantity | 10 g |
| Synonym | 3-Carbomethoxypropanoic acid, 4-Methoxy-4-oxobutanoic Acid, Methyl Hydrogen Succinate, Methyl Monosuccinate, Monomethyl Butanedioate, Monomethyl Succinate NSC 511, 3-(Methoxycarbonyl)propanoic Acid, 3-(Methoxycarbonyl)propionic Acid, Monomethyl Succinic Acid Ester, Methyl Succinic Acid Ester, Monomethyl Butanedioic Acid Ester |
| InChI Formula | InChI=1S/C5H8O4/c1-9-5(8)3-2-4(6)7/h2-3H2,1H3,(H,6,7) |
| SMILES | COC(=O)CCC(=O)O |
| IUPAC Name | 4-methoxy-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 132.11 |
| Formula Weight | 132.0423 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?