missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Succinate, TRC

Product Code. 30481467
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481467 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481467 Supplier Toronto Research Chemicals Supplier No. TRCM5425951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monomethyl Succinate
CAS 3878-55-5
Molecular Formula C5H8O4
Quantity 1 g
Synonym 3-Carbomethoxypropanoic acid, 4-Methoxy-4-oxobutanoic Acid, Methyl Hydrogen Succinate, Methyl Monosuccinate, Monomethyl Butanedioate, Monomethyl Succinate NSC 511, 3-(Methoxycarbonyl)propanoic Acid, 3-(Methoxycarbonyl)propionic Acid, Monomethyl Succinic Acid Ester, Methyl Succinic Acid Ester, Monomethyl Butanedioic Acid Ester
InChI Formula InChI=1S/C5H8O4/c1-9-5(8)3-2-4(6)7/h2-3H2,1H3,(H,6,7)
SMILES COC(=O)CCC(=O)O
IUPAC Name 4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol) 132.11
Formula Weight 132.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.