Learn More
Monopolin Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB500-131DL594
Product Code. 18727840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Monopolin Polyclonal antibody specifically detects Monopolin in Yeast samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monopolin
DyLight 594
RUO
Western Blot
0.1 mL
Store at 4°C in the dark.