missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosialoganglioside GM3 Sodium Salt, TRC
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Specifications
Specifications
| Chemical Name or Material | Monosialoganglioside GM3 Sodium Salt |
| Molecular Formula | C59H107N2NaO21 |
| Quantity | 0.5 mg |
| Synonym | Ganglioside GM3 Sodium Salt, GLac1 Sodium Salt, GM3 Sodium Salt, Ganglioside M3 Sodium Salt, Hematoside Sodium Salt, Sialosyllactosylceramide Sodium Salt |
| InChI Formula | InChI=1S/C59H108N2O21.Na/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-46(69)61-40(41(66)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)38-77-56-51(73)50(72)53(45(37-64)79-56)80-57-52(74)55(49(71)44(36-63)78-57)82-59(58(75)76)34-42(67)47(60-39(3)65)54(81-59)48(70)43(68)35-62;/h30,32,40-45,47-57,62-64,66-68,70-74H,4-29,31,33-38H2,1-3H3,(H,60,65)(H,61,69)(H,75,76);/q;+1/p-1/b32-30+;/t40-,41+,42-,43+,44-,45-,47+,48+,49-,50+,51+,52+,53-,54+,55+,56+,57-,59?;/m0./s1 |
| SMILES | O[C@H]1CC(O[C@H]2[C@@H](O)[C@H](O[C@H]3[C@H](CO)O[C@@H](OC[C@]([C@@](O)([H])/C=C/CCCCCCCCCCCCC)([H])N([H])C(CCCCCCCCCCCCCCCCC)=O)[C@H](O)[C@H]3O)O[C@@H](CO)[C@@H]2O)(C(O[Na])=O)O[C@@H]([C@@]([H])([C@]([H])(O)CO)O)[C@@H]1NC(C)=O |
| Molecular Weight (g/mol) | 1203.47 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction