missing translation for 'onlineSavingsMsg'
Learn More

Montelukast Styrene, TRC

Product Code. 30012025
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012025 1 mg 1mg
30008722 5 mg 5mg
17941345 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012025 Supplier Toronto Research Chemicals Supplier No. TRCM5680301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918972-54-0
Molecular Formula C35 H34 Cl N O2 S
Molecular Weight (g/mol) 568.17
Synonym 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic Acid, Montelukast Styrene, Montekulast Sodium Imp. B (EP)
IUPAC Name 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-(2-prop-1-en-2-ylphenyl)propyl]sulfanylmethyl]cyclopropyl]acetic acid
SMILES CC(=C)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C35H34ClNO2S/c1-24(2)31-9-4-3-7-26(31)13-17-33(40-23-35(18-19-35)22-34(38)39)28-8-5-6-25(20-28)10-15-30-16-12-27-11-14-29(36)21-32(27)37-30/h3-12,14-16,20-21,33H,1,13,17-19,22-23H2,2H3,(H,38,39)/b15-10+/t33-/m1/s1
Formula Weight 567.1999
Chemical Name or Material Montelukast Styrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.