missing translation for 'onlineSavingsMsg'
Learn More

Morantel Tartrate, TRC

Product Code. 30005983
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005983 250 mg 250mg
17991345 100 mg 100mg
17914745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005983 Supplier Toronto Research Chemicals Supplier No. TRCM630350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26155-31-7
Molecular Formula C12 H16 N2 S . C4 H6 O6
Molecular Weight (g/mol) 370.42
Synonym Morantel hydrogen tartrate, 1-Methyl-2-[(E)-2-(3-methylthiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine hydrogen tartrate, Pyrimidine, 1,4,5,6-tetrahydro-1-methyl-2-[(2E)-2-(3-methyl-2-thienyl)ethenyl]-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Pyrimidine, 1,4,5,6-tetrahydro-1-methyl-2-[2-(3-methyl-2-thienyl)ethenyl]-, (E)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), Pyrimidine, 1,4,5,6-tetrahydro-1-methyl-2-[2-(3-methyl-2-thienyl)vinyl]-, (E)-, tartrate (1:1) (8CI), Banminth II, CP 12009-18, Greenguard, Morantel tartrate, Morantrel tartrate, Rumatel, UK 2964-18
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;1-methyl-2-[(E)-2-(3-methylthiophen-2-yl)ethenyl]-5,6-dihydro-4H-pyrimidine
SMILES CN1CCCN=C1\C=C\c2sccc2C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H16N2S.C4H6O6/c1-10-6-9-15-11(10)4-5-12-13-7-3-8-14(12)2;5-1(3(7)8)2(6)4(9)10/h4-6,9H,3,7-8H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/b5-4+;/t;1-,2-/m.1/s1
Formula Weight 370.1199
Percent Purity >95%
Chemical Name or Material Morantel tartrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.