missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MORC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111309
This item is not returnable.
View return policy
Description
Immunogen sequence: SPSLQLKPLD SSVLQFSSKY KWILGEEPVE KRRRLQNEMT TPSLDYSMPA PYRRVEAPVA YPEGENSHDK SSSERSTPPY LFPEYPEASK NTGQNREVSI LYPGAKDQRQ GSLLPEELED Q Highest antigen sequence indentity to the following orthologs: Rat - 64%, Mouse - 66%.
In human, the four current members of the microrchidia (morc) gene family share an N-terminal ATPase-like ATP-binding region and a CW four-cysteine zinc-finger motif. The protein encoded by this gene also has a nuclear matrix binding domain and a two-stranded coiled-coil motif near its C-terminus. This gene is widely expressed at low levels in normal tissues and has elevated expression in placenta and testis. Alternative splicing results in multiple transcript variants encoding distinct proteins.
Specifications
| MORC4 | |
| Polyclonal | |
| Unconjugated | |
| MORC4 | |
| 1600017G11Rik; 5630401M14Rik; AA117449; dJ75H8.2; microrchidia 4; MORC family CW-type zinc finger 4; MORC family CW-type zinc finger protein 4; MORC4; RGD1559905; ZCW4; ZCWCC2; Zinc finger CW-type coiled-coil domain protein 2; zinc finger CW-type domain protein 4; zinc finger, CW type with coiled-coil domain 2; zinc finger, CW-type with coiled-coil domain 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TE76 | |
| MORC4 | |
| Recombinant protein fragment of Human MORC4 (Product #RP-91958) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction