missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORF4 Antibody (OTI5F2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72769PCP
This item is not returnable.
View return policy
Description
MORF4 Monoclonal antibody specifically detects MORF4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MORF4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10934 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5F2 | |
| Western Blot, Immunohistochemistry | |
| CSR, CSRB, mortality factor 4, SEN, SEN1, senescence (cellular)-related 1, senescence-related, cellular, 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-255 of human MORF4 (NP_006783) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction