missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORF4 Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800383
This item is not returnable.
View return policy
Description
Cellular senescence is the terminal non-dividing state that normal cells enter following completion of their proliferative potential. Fusions of immortal human cell lines with each other have led to their assignment to one of four complementation groups. Mortality factor 4 (MORF4) was identified as the lead member of a family of transcription factor-like proteins that reverses this immortal phenotype. Like other members in this family, MORF4 is localized to the nucleus and possesses transcription factor-like motifs such as helix-loop-helix and a leucine zipper motif that might allow it to form transcriptionally active homo- or heterodimers. MORF4 has been shown to bind to the transcription corepressors mSin3A and TLE, suggesting that together, these complexes may play a role in transcriptional repression of genes that lead to cellular senescence.Specifications
| MORF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| CSR, CSRB, SEN, SEN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10934 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1C8 | |
| Unconjugated | |
| MORF4 | |
| MORF4 | |
| Human recombit protein fragment corresponding to amino acids 1-255 of human MORF4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction