missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORF4 Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800383S
This item is not returnable.
View return policy
Description
MORF4 Monoclonal antibody specifically detects MORF4 in Human samples. It is validated for Western BlotSpecifications
| MORF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| 9130413I22Rik; AL022827; csrb; Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; mKIAA4002; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; MORF4 family-associated protein 1; MORF4L1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; mortality factor 4-like protein 1; Mrfap1; mrfap1 {ECO:0000312; MRG15; Pam14; Pgr1; PP368; Protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; protein MSL3-1; RGD:628631}; RGD1306635; S863-6; SEN1; T-cell activation protein; T-cell activation protein-related protein; TEG-189; testis expressed gene 189; testis-expressed gene 189 protein; Tex189; transcription factor-like protein MRG15 | |
| Human recombinant protein fragment corresponding to amino acids 1-255 of human MORF4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| OTI1C8 | |
| Unconjugated | |
| MORF4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10934 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction