missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORN4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 590.10
Specifications
| Antigen | MORN4 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18494682
|
Novus Biologicals
NBP2-30713-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101044
|
Novus Biologicals
NBP2-30713 |
0.1 mL |
€ 624.00 € 590.10 / 0.10mL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MORN4 Polyclonal specifically detects MORN4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MORN4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bA548K23.4, C10orf83, chromosome 10 open reading frame 83, FLJ25925,44050 protein, MORN repeat containing 4, MORN repeat-containing protein 4, Protein 44050, retinophilin | |
| MORN4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8NDC4 | |
| 118812 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FNGVGVFIRYDNMTFEGEFKNGRVDGFGLLTFPDGSHGIPRNEGLFENNKLLRREKCSAIVQRAQSASKSARNL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.