missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOS Antibody (S3.1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2784G
This item is not returnable.
View return policy
Description
MOS Monoclonal specifically detects MOS in Xenopus samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MOS | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MOS | |
| Recombinant (Xenopus laevis) Mos fusion protein | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4342 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| S3.1 | |
| Western Blot, Immunohistochemistry | |
| c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction