missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOSPD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542665
This item is not returnable.
View return policy
Description
Peptide sequence: FLLFLLTGIV SVAFLLLPLP DELGSQLPQV LHVSLGQKLV AAYVLGLLTM Sequence homology: Cow: 92%; Dog: 92%; Goat: 77%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.
MOSPD3 is a multi-pass membrane protein with a major sperm protein (MSP) domain. The deletion of a similar mouse gene is associated with defective cardiac development and neonatal lethality.This gene encodes a multi-pass membrane protein with a major sperm protein (MSP) domain. The deletion of a similar mouse gene is associated with defective cardiac development and neonatal lethality. Alternate transcriptional splice variants, encoding different isoforms, have been described.
Specifications
| MOSPD3 | |
| Polyclonal | |
| Unconjugated | |
| MOSPD3 | |
| 1190005J19Rik; 5133401H10Rik; CDS3; gene trap integration gene 2; Gtig2; hypothetical protein LOC506163; Mospd3; motile sperm domain containing 3; motile sperm domain-containing protein 3; NET30; R124 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64598 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75425 | |
| MOSPD3 | |
| Synthetic peptide directed towards the C-terminal of human MOSPD3 (aa 181-230). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu