Learn More
Jenway™ Moulded Cuvette Rack
Product Code. 11434839
Click to view available options
Holds:
Cuvettes (10 x 10 mm)
Unit Size:
Each
Product Code. 11434839
Brand: Jenway™ 9961035
Only null left
Add to basket
Add to basket
Holds 16 Cuvettes, 10 x 10mm
For use with Jenway 63 and 67 Series Spectrophotmeters
Specifications
Holds Cuvettes (10 x 10 mm)
Positions 16
For Use With (Equipment) Jenway 63 and 67 Series Spectrophotmeters