Learn More
Ibidi Mounting Medium with DAPI
Product Code. 17246423
Encompass_Preferred
Click to view available options
Product Type:
Mounting Medium
Unit Size:
Each
Product Code. 17246423
Brand: Ibidi 50011
Only null left
Add to basket
Add to basket
Ibidi Mounting Medium with DAPI is a mounting medium with DAPI that is optimized for fluorescence microscopy.
  • A mounting medium with DAPI that is optimized for fluorescence microscopy with Ibidi μ-Slides and μ-Dishes
  • non-drying
  • non-hardening
  • volume stable
  • 15 mL
Specifications
Quantity 1
Product Type Mounting Medium
Volume (Metric) 5 mL