missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 4-1BB Ligand/TNFSF9 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1246-500
This item is not returnable.
View return policy
Description
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| 4-1BB Ligand/TNFSF9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P41274 | |
| TNFSF9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9 Arg104-Glu309 Accession # P41274 | |
| 500 μg | |
| Primary | |
| Detects mouse 4-1BB Ligand/TNFSF9 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human RELT is observed. | |
| Mouse | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 203942 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8744 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction