missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 5'-Nucleotidase/CD73 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4488-SP
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| 5'-Nucleotidase/CD73 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q61503 | |
| NT5E | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse 5'-Nucleotidase/CD73 Trp29-Lys549 Accession # Q61503 | |
| 25 μg | |
| Primary | |
| Detects mouse 5'-Nucleotidase/CD73 in direct ELISAs. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 496406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5' nucleotidase (CD73), 5'-NT, 5-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NT5E, NTE, Purine 5-Prime-Nucleotidase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4907 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction