missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Activin C beta C subunit Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF489
This item is not returnable.
View return policy
Description
Activin C/Inhibin beta C Polyclonal specifically detects Activin C/Inhibin beta C in Mouse samples. It is validated for Western Blot.
Specifications
| Activin C/Inhibin beta C | |
| Polyclonal | |
| Unconjugated | |
| inhibin beta C chain, IHBC, INHBC, inhibin, beta C | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3626 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| INHBC | |
| E. coli-derived recombinant mouse Activin C peptide aa 236-352 | |
| 100 μg | |
| Primary | |
| Detects mouse Activin C beta C subunit in direct ELISAs and Western blots. In direct ELISA, approximately 5% cross-reactivity with recombinant human (rh) Activin A and rhInhibin A is observed and no cross-reactivity with rhInhibin B is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction