missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse AHR Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF6697-SP
This item is not returnable.
View return policy
Description
AHR Polyclonal specifically detects AHR in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| AHR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 196 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P30561 | |
| AHR | |
| E. coli-derived recombinant mouse AHR Asn706-Ser805 (Thr758Ala) Accession # P30561 | |
| 25 μg | |
| Primary | |
| Detects mouse AHR in Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction