Learn More
A4GALT, Mouse, Clone: 3E12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant A4GALT.
Brand: Abnova H00053947-M08A.200uL
Description
The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor. [provided by RefSeq
Sequence: LTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVHVWNKKSQGTRFEATSRALLAQLHARYCPTTHEAMKMYLSpecifications
| A4GALT | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_017436 | |
| A4GALT | |
| A4GALT (NP_059132,254 a.a. ∽ 353 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 53947 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 3E12 | |
| Mouse monoclonal antibody raised against a partial recombinant A4GALT. | |
| A4GALT | |
| A14GALT/A4GALT1/P1/PK | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.