Learn More
AATK, Mouse, Clone: 5B8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AATK.
Brand: Abnova H00009625-M03A.200uL
Description
The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line. [provided by RefSeq
Sequence: SPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSAYFSDLEAEAEATSGPEKKCGGDRAPGPELGLRSTGQPSEQVCLRPGVSGSpecifications
| AATK | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC047378 | |
| AATK | |
| AATK (AAH47378,161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9625 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5B8 | |
| Mouse monoclonal antibody raised against a partial recombinant AATK. | |
| AATK | |
| AATYK/AATYK1/KIAA0641/LMR1/LMTK1/p35BP | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.