Learn More
ADAM17, Mouse, Clone: 1F6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADAM17.
Brand: Abnova H00006868-M01A.200uL
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biologic processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene functions as a tumor necrosis factor-alpha converting enzyme; binds mitotic arrest deficient 2 protein; and also plays a prominent role in the activation of the Notch signaling pathway. [provided by RefSeq
Sequence: RADPDPMKNTCKLLVVADHRFYRYMGRGEESTTTNYLIELIDRVDDIYRNTSWDNAGFKGYGIQIEQIRILKSPQEVKPGEKHYNMAKSYPNEEKDAWDVSpecifications
| ADAM17 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_003183.4 | |
| ADAM17 | |
| ADAM17 (NP_003174.3,215 a.a. ∽ 314 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6868 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1F6 | |
| Mouse monoclonal antibody raised against a partial recombinant ADAM17. | |
| ADAM17 | |
| CD156b/MGC71942/TACE/cSVP | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.