missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin, Mouse, Clone: Adn214, Abnova™
Mouse monoclonal antibody raised against native adiponectin.
Brand: Abnova MAB8467.1mg
This item is not returnable.
View return policy
Specifications
| Adiponectin | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| Adn214 | |
| Mouse monoclonal antibody raised against native adiponectin. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Native purified adiponectin from human serum. | |
| RUO | |
| Recommended pairs for low molecular weight adiponectin immunodetection in sandwich immunoassay (capture-detection) : Adn214-Adn27. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido