missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin, Mouse, Clone: Adn97, Abnova™
Mouse monoclonal antibody raised against native adiponectin.
Brand: Abnova MAB8465.1mg
This item is not returnable.
View return policy
Specifications
| Adiponectin | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| Adn97 | |
| Mouse monoclonal antibody raised against native adiponectin. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Native purified adiponectin from human serum. | |
| RUO | |
| Recommended pairs for total adiponectin immunodetection in sandwich immunoassay (capture-detection) : Adn97-Adn94. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction