Learn More
AFAP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human AFAP1 protein.
Brand: Abnova H00060312-B01P.50ug
Description
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MEELIVELRLFLELLDHEYLTSTVREKKAVITNILLRIQSSKGFDVKDHAQKQETANSLPAPPQMPLPEIPQPWLPPDSGPPPLPTSSLPEGYYEEAVPLSPGKAPEYITSNYDSDAMSSSYESYDEEEEDGKGKKTRHQWPSEEASMDLVKDAKICAFLLRKKRFGQWTKLLCVIKDTKLLCYKSSKDQQPQMELPLQGCNITYIPKDSKKKKHELKITQQGTDPLVLAVQSKEQAEQWLKVIKEAYSGCSGPVDSECPPPPSSPVHKAELEKKLSSERPSSDGEGVVENGITTCNGKEQVKRKKSSKSEAKGTVSKVTGKKITKIISLGKKKPSTDEQTSSAEEDVPTCGYLNVLSNSRWRERWCRVKDNKLIFHKDRTDLKTHIVSIPLRGCEVIPGLDCKHPLTFRLLRNGQEVAVLEASSSEDMGRWIGILLAETGSSTDPEALHYDYIDVEMSASVIQTAKQTFCFMNRRVISANPYLGGTSNGYAHPSGTALHYDDVPCINGSLKGKKPPVASNGVTGKGKTLSSQPKKADPAAVVKRTGSNAAQYKYGKNRVEADAKRLQTKEEELLKRKEALRNRLAQLRKERKDLRAAIEVNAGRKPQAILEEKLKQLEEECRQKEAERVSLELELTEVKESLKKALAGGVTLGLAIEPKSGTSSPQSPVFRHRTLENSPISSCDTSDTEGPVPVNSAAVLKKSQAAPGSSPCRGHVLRKAKEWELKNGTSpecifications
| AFAP1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human AFAP1 protein. | |
| AFAP1 | |
| AFAP/AFAP-110/FLJ56849 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 60312 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032777.1 | |
| AFAP1 | |
| AFAP1 (AAH32777.1,1 a.a. ∽ 730 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only