Learn More
AKR1B1, Mouse, Clone: 19, Abnova™
Mouse monoclonal antibody raised against partial recombinant AKR1B1.
Brand: Abnova MAB9983.100ug
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member catalyzes the reduction of a number of aldehydes, including the aldehyde form of glucose, and is thereby implicated in the development of diabetic complications by catalyzing the reduction of glucose to sorbitol. Multiple pseudogenes have been identified for this gene. The nomenclature system used by the HUGO Gene Nomenclature Committee to define human aldo-keto reductase family members is known to differ from that used by the Mouse Genome Informatics database. [provided by RefSeq
Specifications
| AKR1B1 | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| AKR1B1 | |
| AKR1B1 | |
| Recombinant protein corresponding to amino acids 33-316 of human AKR1B1. | |
| 100 μg | |
| Primary | |
| It can expression in Jurkat whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| ELISA, Western Blot | |
| 19 | |
| Mouse monoclonal antibody raised against partial recombinant AKR1B1. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| ADR/ALDR1/ALR2/AR/MGC1804 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 231 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.