Learn More
AOF2, Mouse, Clone: 1B2E5, Abnova™
Mouse monoclonal antibody raised against partial recombinant AOF2.
Brand: Abnova MAB5530.100ug
Description
This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| AOF2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:2000) Immunohistochemistry (Frozen sections) (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end | |
| AOF2 | |
| AOF2 | |
| Recombinant protein corresponding to human AOF2. | |
| RUO | |
| 23028 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 1B2E5 | |
| Mouse monoclonal antibody raised against partial recombinant AOF2. | |
| In ascites | |
| BHC110/KDM1/KIAA0601/LSD1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.