Learn More
AP4M1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AP4M1.
Brand: Abnova H00009179-A01.50uL
Description
This gene encodes a subunit of the heterotetrameric AP-4 complex. The encoded protein belongs to the adaptor complexes medium subunits family. This AP-4 complex is involved in the recognition and sorting of cargo proteins with tyrosine-based motifs from the trans-golgi network to the endosomal-lysosomal system. [provided by RefSeq
Sequence: AELAEGALRWDLPRVQGGSQLSGLFQMDVPGPPGPPSHGLSTSASPLGLGPASLSFELPRHTCSGLQVRFLRLAFRPCGNANPHKWVRHLSHSDAYVIRISpecifications
| AP4M1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant AP4M1. | |
| AP4M1 | |
| MU-4/MU-ARP2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004722 | |
| AP4M1 | |
| AP4M1 (NP_004713, 354 a.a. to 453 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9179 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only