Learn More
ARF1, Mouse, Clone: AT1B3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant ARF1.
Brand: Abnova MAB10011.100uL
Description
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| ARF1 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against full length recombinant ARF1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| ARF1 | |
| Recombinant protein corresponding to full length human ARF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| AT1B3 | |
| Unconjugated | |
| ELISA Flow Cytometry (2.5 ug for 106 cells) Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ARF1 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 375 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.