missing translation for 'onlineSavingsMsg'
Learn More

ATP12A, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16165024
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16165024 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16165024 Supplier Abnova Supplier No. H00000479A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant ATP12A.

The protein encoded by this gene belongs to the family of P-type cation transport ATPases. This gene encodes a catalytic subunit of the ouabain-sensitive H+/K+ -ATPase that catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. It is also responsible for potassium absorption in various tissues. [provided by RefSeq]

Sequence: SSFNKMIPQQALVIRDSEKKTIPSEQLVVGDIVEVKGGDQIPADIRVLSSQGCRVDNSSLTGESEPQPRSSEFTHENPLETKNICFYSTTCLEASTSPVGTVTGMVIN

Specifications

Antigen ATP12A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ATP12A.
Formulation 50% glycerol
Gene ATP12A
Gene Accession No. NM_001676
Gene Alias ATP1AL1
Gene Symbols ATP12A
Host Species Mouse
Immunogen ATP12A (NP_001667, 174 a.a. to 281 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 479
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.