Learn More
B2M, Mouse, Clone: D6D1, Abnova™
Mouse monoclonal antibody raised against recombinant B2M.
Brand: Abnova MAB8112.500ug
Description
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia
Specifications
| B2M | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| B2M | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA | |
| D6D1 | |
| Mouse monoclonal antibody raised against recombinant B2M. | |
| Lyophilized from 10mM PBS | |
| B2M | |
| Recombinant protein corresponding to human B2M. | |
| RUO | |
| 567 | |
| Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.