Learn More
B3GAT3, Mouse, Clone: 4C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant B3GAT3.
Brand: Abnova H00026229-M01A.200uL
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans. [provided by RefSeq
Sequence: VVGFHTAWEPSRPFPVDMAGFAVALPLLLDKPNAQFDSTAPRGHLESSLLSHLVDPKDLEPRAANCTRVLVWHTRTEKPKMKQEEQLQRQGRGSDPAIEVSpecifications
| B3GAT3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_012200 | |
| B3GAT3 | |
| B3GAT3 (NP_036332,236 a.a. ∽ 335 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 26229 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4C12 | |
| Mouse monoclonal antibody raised against a partial recombinant B3GAT3. | |
| B3GAT3 | |
| GLCATI/GlcAT-I | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.