Learn More
BAG2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BAG2.
Brand: Abnova H00009532-A01.50uL
Description
BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner. [provided by RefSeq
Sequence: RNPQQQESLKHATRIIDEVVNKFLDDLGNAKSHLMSLYSACSSEVPHGPVDQKFQSIVIGCALEDQKKIKRRLETLLRNIENSDKAIKLLEHSKGAGSKTLQQNAESRFNSpecifications
| BAG2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant BAG2. | |
| BAG2 | |
| BAG-2/KIAA0576/MGC149462/dJ417I1.2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| NM_004282 | |
| BAG2 | |
| BAG2 (NP_004273, 102 a.a. to 211 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9532 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only