missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBC3, Mouse, Clone: 10D4G7, Abnova™
Mouse monoclonal antibody raised against recombinant BBC3.
Brand: Abnova MAB2757.100ug
This item is not returnable.
View return policy
Specifications
| BBC3 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| BBC3 | |
| BBC3 | |
| Recombinant protein corresponding to amino acids 76-170 of human BBC3. | |
| RUO | |
| 27113 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Western Blot | |
| 10D4G7 | |
| Mouse monoclonal antibody raised against recombinant BBC3. | |
| In PBS (0.02% sodium azide) | |
| JFY1/PUMA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction