missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS4, Mouse, Clone: 4D4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BCAS4.
Brand: Abnova H00055653-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MLLVDADQPEPMRSGARELALFLTPEPGAEAKEVEETIEGMLLRLEEFCSLADLIRSDTSQILEENIPVLKAKLTEMRGIYAKVDRLEAFVKMVGHHVAFLEADVLQAERDHGAFPQALRRWLGSAGLPSFRNKSPAPVPVTYELPTLYRTEDYFPVDAGEAQHHPRTCPRPLSpecifications
| BCAS4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC038381.1 | |
| BCAS4 | |
| BCAS4 (AAH38381.1,1 a.a. ∽ 173 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4D4 | |
| Mouse monoclonal antibody raised against a full-length recombinant BCAS4. | |
| BCAS4 | |
| CNOL/FLJ20495 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 55653 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction