missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2, Mouse, Clone: 10C4, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of BCL2.
Brand: Abnova MAB1794.100ug
This item is not returnable.
View return policy
Specifications
| BCL2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Bcl2 | |
| Bcl2 | |
| A synthetic peptide corresponding to amino acids 61-76 of mouse BCL2. | |
| RUO | |
| 12043 | |
| Liquid |
| Western Blot | |
| 10C4 | |
| Mouse monoclonal antibody raised against synthetic peptide of BCL2. | |
| In PBS (0.08% sodium azide) | |
| AW986256/Bcl-2/C430015F12Rik/D630044D05Rik/D830018M01Rik | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction