Learn More
BCL2, Mouse, Clone: 7, Abnova™
Mouse monoclonal antibody raised against partial recombinant BCL2.
Brand: Abnova MAB9891.100ug
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Specifications
| BCL2 | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| BCL2 | |
| Bcl-2 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 596 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 7 | |
| Mouse monoclonal antibody raised against partial recombinant BCL2. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| NM_000633 | |
| BCL2 | |
| Recombinant protein corresponding to amino acids 1-195 of human BCL2. | |
| 100 μg | |
| Primary | |
| It can expression in Jurkat whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.