Learn More
BCL2, Mouse, Clone: 8C8, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of BCL2.
Brand: Abnova MAB11265.100ug
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Sequence: GAAPAPGIFSSQPGSpecifications
| BCL2 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) Western Blot (1-3 ug/ml) The optimal working dilution should be determined by the end user. | |
| BCL2 | |
| BCL2 | |
| A synthetic peptide corresponding to amino acids 41-54 of human BCL2. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 8C8 | |
| Mouse monoclonal antibody raised against synthetic peptide of BCL2. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| Bcl-2 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 596 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.