Learn More
BCL2, Mouse, Clone: AT1B5, Abnova™
Mouse monoclonal antibody raised against partial recombinant BCL2.
Brand: Abnova MAB9719.100uL
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Specifications
| BCL2 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant BCL2. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Bcl-2 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 596 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| ELISA, Immunofluorescence, Western Blot | |
| AT1B5 | |
| Unconjugated | |
| ELISA (1:500) Western Blot (1:250-1:500) Immunofluorescence (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| BCL2 | |
| BCL2 | |
| Recombinant protein corresponding to amino acids 1-211 of human Bcl-2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.