missing translation for 'onlineSavingsMsg'
Learn More

beta 2 (MG), Mouse, Clone: D6E4, Abnova™

Product Code. 16201570
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16201570 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16201570 Supplier Abnova Supplier No. MAB6375.500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant beta 2 (MG).

Specifications

Antigen beta 2 (MG)
Applications ELISA
Classification Monoclonal
Clone D6E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant beta 2 (MG).
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen Recombinant protein corresponding to human beta 2 Micro Globulin (MG).
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.