missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin, Mouse, Clone: SB58c, Abnova™
Mouse monoclonal antibody raised against BIOT.
Brand: Abnova MAB3274.500ug
This item is not returnable.
View return policy
Specifications
| Biotin | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1 ug/106 cells) ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μg | |
| Primary | |
| Store at 4°C. | |
| IgG1 |
| ELISA, Flow Cytometry | |
| SB58c | |
| Mouse monoclonal antibody raised against BIOT. | |
| In 100mM BBS, pH 8.2 | |
| KLH1139-biotin. | |
| RUO | |
| Biotin. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction