missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin, Mouse, HRP, Clone: SB58c, Abnova™
Mouse monoclonal antibody raised against BIOT.
Brand: Abnova MAB9877.1mL
This item is not returnable.
View return policy
Specifications
| Biotin | |
| Monoclonal | |
| HRP | |
| Immunofluorescence (1:4000-1:8000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mL | |
| Primary | |
| Liquid |
| ELISA | |
| SB58c | |
| Mouse monoclonal antibody raised against BIOT. | |
| In PBS, pH 7.4 (50% glycerol) | |
| KLH1139-biotin. | |
| RUO | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction