missing translation for 'onlineSavingsMsg'
Learn More

BRIP1, Mouse, Clone: pp15-IB4, Abnova™

Product Code. 16095636
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16095636 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16095636 Supplier Abnova Supplier No. MAB2402.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against synthetic peptide of BRIP1.

The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations. [provided by RefSeq

Specifications

Antigen BRIP1
Applications Western Blot
Classification Monoclonal
Clone pp15-IB4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of BRIP1.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene BRIP1
Gene Alias BACH1/FANCJ/FLJ90232/MGC126521/MGC126523/OF
Gene Symbols BRIP1
Host Species Mouse
Immunogen A synthetic peptide corresponding to human BRIP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83990
Test Specificity This antibody is specific to Brip1/FANCJ.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.