missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine, Mouse, Clone: SPM166, Abnova™
Mouse monoclonal antibody raised against Bromodeoxyuridine.
Brand: Abnova MAB12072.100ug
This item is not returnable.
View return policy
Specifications
| Bromodeoxyuridine | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| SPM166 | |
| Mouse monoclonal antibody raised against Bromodeoxyuridine. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| Bromodeoxyuridine (BrdU) conjugated to KLH. | |
| 100 μg | |
| Primary | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction