Learn More
BUB1, Mouse, Clone: P8.14H5.1H1, Abnova™
Mouse monoclonal antibody raised against BUB1.
Brand: Abnova MAB8963.100ug
Description
This gene encodes a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint. Mutations in this gene have been associated with aneuploidy and several forms of cancer. [provided by RefSeq
Specifications
| BUB1 | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against BUB1. | |
| In PBS, pH 7.4 (0.02% sodium azide) | |
| BUB1A/BUB1L/hBUB1 | |
| Mouse | |
| Purified ascites | |
| RUO | |
| 699 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| P8.14H5.1H1 | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| BUB1 | |
| BUB1 | |
| Recombinant GST fusion protein corresponding to C-terminus (642 amino acids) of human BUB1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.