missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacna1g, Mouse, ATTO488, Clone: S178A-9, Abnova™
Mouse monoclonal antibody raised against recombinant of Cacna1g.
Brand: Abnova MAB11550.100ug
This item is not returnable.
View return policy
Specifications
| Cacna1g | |
| Monoclonal | |
| Atto 488 | |
| Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Cacna1g | |
| Cacna1g | |
| Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g. | |
| 100 μg | |
| Primary | |
| Detects ∼<200 kDa. Does not cross-react with Cacna1h. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunohistochemistry, Western Blot | |
| S178A-9 | |
| Mouse monoclonal antibody raised against recombinant of Cacna1g. | |
| In PBS, pH 7.4 (50% glycerol) | |
| Cav3.1d/[a]1G/a1G/alpha-1G/mKIAA1123 | |
| Mouse | |
| Protein G Purified | |
| RUO | |
| 12291 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction