missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALCA, Mouse, Clone: 3B1 1C10, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CALCA.
Brand: Abnova MAB3673.1mg
This item is not returnable.
View return policy
Description
Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes reduction in serum calcium, an effect opposite to that of parathyroid hormone (PTH; MIM 168450).[supplied by OMIM
Specifications
| CALCA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CALCA | |
| CALCA | |
| A synthetic peptide corresponding to human CALCA. | |
| RUO | |
| 796 | |
| Human | |
| Liquid |
| ELISA | |
| 3B1 1C10 | |
| Mouse monoclonal antibody raised against synthetic peptide of CALCA. | |
| In BBS | |
| CALC1/CGRP/CGRP-I/CGRP1/CT/KC/MGC126648 | |
| Mouse | |
| 1 mg | |
| Primary | |
| This antibody recognizes natural human CALCA. | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction