missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine parvovirus, Mouse, Clone: 8H7, Abnova™
Mouse monoclonal antibody raised against canine parvovirus.
Brand: Abnova MAB0590.200ug
This item is not returnable.
View return policy
Specifications
| Canine parvovirus | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at 4°C. | |
| IgG2a |
| ELISA, Inhibition Assays | |
| 8H7 | |
| Mouse monoclonal antibody raised against canine parvovirus. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Canine parvovirus. | |
| RUO | |
| Virus | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction