Learn More
CCL27/CTACK Mouse anti-Human, Clone: 124308, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB376-100
Encompass
Product Code. 15736474
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.