Learn More
CD14, Mouse, Clone: MEM-15, Abnova™
Mouse monoclonal antibody raised against full-length native CD14.
Brand: Abnova MAB3831.100ug
Description
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq
Specifications
| CD14 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD14 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with CD14, a 53-55 KDa GPI (glycosylphosphatidylinositol)-linked membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. This antibody MEM-15 also reacts with soluble forms of CD14 found in serum and in the urine of some nephrotic patients. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunoprecipitation | |
| MEM-15 | |
| Mouse monoclonal antibody raised against full length native CD14. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD14 | |
| Native purified human CD14. | |
| RUO | |
| 929 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.